Assay Details
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003182.1 | NP_001003182.1 | 3 - 4 | 246 | 63 | |
XM_022415492.1 | XP_022271200.1 | 3 - 4 | 364 | 63 | ||
GenBank mRNA | AB016817.1 | - | 3 - 4 | 236 | 63 | |
AF077817.1 | - | 3 - 4 | 246 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
