Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618833.2 | XP_005618890.1 | 923 | 69 | ||
| GenBank mRNA | AB023422.1 | - | 669 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618833.2 | XP_005618890.1 | 923 | 69 | ||
| GenBank mRNA | AB023422.1 | - | 669 | 69 | ||
Set Membership: |
|