Assay Details
| Gene Symbol: CYP21A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003335.1 | NP_001003335.1 | 6 - 7 | 790 | 89 | |
| GenBank mRNA | AB039881.1 | - | 6 - 7 | 790 | 89 | |
More Information
Set Membership: |
|

CYP21A2
Genomic Map