Assay Details
| Gene Symbol: CYP21A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003335.1 | NP_001003335.1 | 9 - 10 | 1277 | 71 | |
| GenBank mRNA | AB039881.1 | - | 9 - 10 | 1277 | 71 | |
More Information
Set Membership: |
|

CYP21A2
Genomic Map