Assay Details
| Gene Symbol: GHRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003052.2 | NP_001003052.1 | 244 | 63 | ||
| XM_005632010.3 | XP_005632067.1 | 277 | 63 | |||
| GenBank mRNA | AB060700.1 | - | 244 | 63 | ||
| AJ298295.1 | - | 266 | 63 | |||
| AJ298296.1 | - | 253 | 63 | |||
More Information
Set Membership: |
|

GHRL
Genomic Map