Assay Details
| Gene Symbol: CCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003010.2 | NP_001003010.1 | 2 - 3 | 236 | 69 | |
| GenBank mRNA | AB098562.1 | - | 2 - 3 | 239 | 69 | |
More Information
Set Membership: |
|

CCL5
Genomic Map