Assay Details
| Gene Symbol: AICDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003380.1 | NP_001003380.1 | 4 - 5 | 623 | 75 | |
| XM_022410855.1 | XP_022266563.1 | 3 - 4 | 677 | 75 | ||
| GenBank mRNA | AB122019.1 | - | 4 - 5 | 623 | 75 | |
More Information
Set Membership: |
|

AICDA
Genomic Map