Assay Details
| Gene Symbol: CCL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005256.1 | NP_001005256.1 | 2 - 3 | 189 | 75 | |
| GenBank mRNA | AB163919.1 | - | 2 - 3 | 189 | 75 | |
More Information
Set Membership: |
|

CCL19
Genomic Map