Assay Details
| Gene Symbol: CANF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003189.2 | NP_001003189.1 | 1 - 2 | 168 | 134 | |
| XM_022422533.1 | XP_022278241.1 | 1 - 2 | 300 | 134 | ||
More Information
Set Membership: |
|

CANF2
Genomic Map