Assay Details
| Gene Symbol: PDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003076.1 | NP_001003076.1 | 3 - 4 | 307 | 140 | |
| GenBank mRNA | AF046874.1 | - | 3 - 4 | 307 | 140 | |
| Y17697.1 | - | 296 | 140 | |||
More Information
Set Membership: |
|

PDC
Genomic Map