Assay Details
| Gene Symbol: IL2RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003211.2 | NP_001003211.2 | 410 | 101 | ||
| XM_005617076.3 | XP_005617133.1 | 2405 | 101 | |||
| XM_022405275.1 | XP_022260983.1 | 559 | 101 | |||
| GenBank mRNA | AB501356.1 | - | 410 | 101 | ||
| AF056491.1 | - | 360 | 101 | |||
| GU207463.1 | - | 360 | 101 | |||
More Information
Set Membership: |
|

IL2RA
Genomic Map