Assay Details
| Gene Symbol: SLC3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003109.1 | NP_001003109.1 | 7 - 8 | 1449 | 71 | |
| XM_022423985.1 | XP_022279693.1 | 8 - 9 | 1533 | 71 | ||
| GenBank mRNA | AF187966.1 | - | 7 - 8 | 1449 | 71 | |
More Information
Set Membership: |
|

SLC3A1
Genomic Map