Assay Details
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003121.1 | NP_001003121.1 | 2 - 3 | 734 | 139 | |
GenBank mRNA | AF206513.1 | - | 2 - 3 | 734 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
