Assay Details
| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415693.1 | XP_022271401.1 | 9 - 10 | 1201 | 82 | |
| XM_538150.6 | XP_538150.3 | 9 - 10 | 1203 | 82 | ||
| GenBank mRNA | AF314532.1 | - | 8 - 9 | 1042 | 82 | |
More Information
Set Membership: |
|

IL13RA1
Genomic Map