Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003169.1 | NP_001003169.1 | 224 | 111 | ||
| XM_005619483.3 | XP_005619540.1 | 389 | 111 | |||
| XM_005619484.3 | XP_005619541.1 | 400 | 111 | |||
| GenBank mRNA | AF327900.1 | - | 129 | 111 | ||
| Y11133.1 | - | 224 | 111 | |||
More Information
Set Membership: |
|

IL18
Genomic Map