Assay Details
| Gene Symbol: NEFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003352.1 | NP_001003352.1 | 2 - 3 | 1144 | 63 | |
| XM_005636006.3 | XP_005636063.1 | 2 - 3 | 1147 | 63 | ||
| XM_005636008.3 | XP_005636065.1 | 2 - 3 | 1147 | 63 | ||
| XM_005636009.3 | XP_005636066.1 | 2 - 3 | 1147 | 63 | ||
| XM_005636010.3 | XP_005636067.1 | 2 - 3 | 1147 | 63 | ||
| XM_014107712.2 | XP_013963187.1 | 2 - 3 | 1147 | 63 | ||
| GenBank mRNA | AF346625.1 | - | 2 - 3 | 1144 | 63 | |
More Information
Set Membership: |
|

NEFH
Genomic Map