Assay Details
| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003063.1 | NP_001003063.1 | 4 - 5 | 581 | 70 | |
| GenBank mRNA | AF448499.1 | - | 4 - 5 | 581 | 70 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPP2CA
Genomic Map