Assay Details
| Gene Symbol: CASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195151.1 | NP_001182080.1 | 822 | 72 | ||
| XM_005629484.3 | XP_005629541.1 | 843 | 72 | |||
| XM_022403604.1 | XP_022259312.1 | 1013 | 72 | |||
| XM_022403606.1 | XP_022259314.1 | 992 | 72 | |||
| GenBank mRNA | AF462046.1 | - | 472 | 72 | ||
More Information
Set Membership: |
|

CASP2
Genomic Map