Assay Details
| Gene Symbol: IL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197188.1 | NP_001184117.1 | 1 | 345 | 80 | |
| XM_022405861.1 | XP_022261569.1 | 369 | 80 | |||
| GenBank mRNA | AF479882.1 | - | 1 | 108 | 80 | |
| AY456092.1 | - | 1 | 103 | 80 | ||
| DN269137.1 | - | 346 | 80 | |||
| DN879783.1 | - | 1 | 300 | 80 | ||
More Information
Set Membership: |
|

IL15
Genomic Map