Assay Details
| Gene Symbol: RPS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313783.1 | NP_001300712.1 | 3 - 4 | 438 | 93 | |
| GenBank mRNA | AJ388520.1 | - | 1 - 2 | 79 | 93 | |
More Information
Set Membership: |
|

RPS11
Genomic Map