Assay Details
| Gene Symbol: SPINT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286963.1 | NP_001273892.1 | 1 - 2 | 411 | 57 | |
| XM_022410678.1 | XP_022266386.1 | 1 - 2 | 450 | 57 | ||
| GenBank mRNA | AJ388543.1 | - | 1 - 2 | 411 | 57 | |
| DN350608.1 | - | 1 - 2 | 115 | 57 | ||
More Information
Set Membership: |
|

SPINT2
Genomic Map