Assay Details
| Gene Symbol: FABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287051.1 | NP_001273980.1 | 2 - 3 | 369 | 69 | |
| GenBank mRNA | AY058916.1 | - | 1 - 2 | 172 | 69 | |
More Information
Set Membership: |
|

FABP1
Genomic Map