Assay Details
| Gene Symbol: PIGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287152.1 | NP_001274081.1 | 1219 | 70 | ||
| GenBank mRNA | AY081057.1 | - | 871 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PIGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287152.1 | NP_001274081.1 | 1219 | 70 | ||
| GenBank mRNA | AY081057.1 | - | 871 | 70 | ||
Set Membership: |
|