Assay Details
Gene Symbol: SSTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623953.3 | XP_005624010.1 | 1158 | 68 | ||
XM_005623955.3 | XP_005624012.1 | 1158 | 68 | |||
XM_014116187.2 | XP_013971662.1 | 1311 | 68 | |||
GenBank mRNA | AY305401.1 | - | 423 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
