Assay Details
Gene Symbol: SSTR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286852.1 | NP_001273781.1 | 1 | 100 | ||
GenBank mRNA | AY305403.1 | - | 1 | 136 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
