Assay Details
| Gene Symbol: P2RX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_540614.4 | XP_540614.1 | 7 - 8 | 1755 | 86 | |
| GenBank mRNA | AY708378.1 | - | 1 - 2 | 98 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: P2RX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_540614.4 | XP_540614.1 | 7 - 8 | 1755 | 86 | |
| GenBank mRNA | AY708378.1 | - | 1 - 2 | 98 | 86 | |
Set Membership: |
|