Assay Details
| Gene Symbol: CXCL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128097.1 | NP_001121569.1 | 188 | 86 | ||
| NM_001308461.1 | NP_001295390.1 | 1 - 2 | 188 | 86 | ||
| GenBank mRNA | DN271893.1 | - | 1 - 2 | 214 | 86 | |
| DN880217.1 | - | 1 - 2 | 80 | 86 | ||
| DQ182700.1 | - | 188 | 86 | |||
| DQ182701.1 | - | 188 | 86 | |||
More Information
Set Membership: |
|

CXCL12
Genomic Map