Assay Details
Gene Symbol: DEFB122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132650.1 | 2 - 3 | 330 | 68 | ||
GenBank mRNA | AY169791.1 | - | 2 - 3 | 253 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
