Assay Details
Gene Symbol: KLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002986.1 | NP_001002986.1 | 5 - 6 | 719 | 149 | |
GenBank mRNA | AY191818.1 | - | 5 - 6 | 719 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() |
