Assay Details
| Gene Symbol: AKR1C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012344.1 | NP_001012344.1 | 45 | 77 | ||
| GenBank mRNA | AY875970.1 | - | 45 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001012344.1 transcripts.
Set Membership: |
|

AKR1C3
Genomic Map