Assay Details
| Gene Symbol: FCER1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110766.2 | NP_001104236.2 | 3 - 4 | 332 | 63 | |
| XM_022415289.1 | XP_022270997.1 | 3 - 4 | 388 | 63 | ||
| GenBank mRNA | D16413.1 | - | 3 - 4 | 340 | 63 | |
More Information
Set Membership: |
|

FCER1A
Genomic Map