Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031820.1 | NP_001026990.1 | 1 - 2 | 192 | 103 | |
XM_005631061.3 | XP_005631118.1 | 2 - 3 | 339 | 103 | ||
XM_005631062.2 | XP_005631119.1 | 2 - 3 | 337 | 103 | ||
XM_005631063.2 | XP_005631120.1 | 2 - 3 | 328 | 103 | ||
XM_022405079.1 | XP_022260787.1 | 2 - 3 | 217 | 103 | ||
GenBank mRNA | DQ127247.1 | - | 1 - 2 | 192 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
