Assay Details
| Gene Symbol: PEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048092.1 | NP_001041557.1 | 3 - 4 | 344 | 90 | |
| GenBank mRNA | DQ130016.1 | - | 3 - 4 | 344 | 90 | |
More Information
Set Membership: |
|

PEBP1
Genomic Map