Assay Details
| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038666.1 | NP_001033755.1 | 6 - 7 | 691 | 123 | |
| XM_022406246.1 | XP_022261954.1 | 6 - 7 | 724 | 123 | ||
| GenBank mRNA | DQ227277.1 | - | 6 - 7 | 691 | 123 | |
More Information
Set Membership: |
|

ADGRE3
Genomic Map