Assay Details
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197043.1 | NP_001183972.1 | 4 - 5 | 718 | 78 | |
XM_005623184.3 | XP_005623241.1 | 5 - 6 | 854 | 78 | ||
GenBank mRNA | HQ637386.1 | - | 4 - 5 | 713 | 78 | |
L23429.1 | - | 3 - 4 | 707 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
