Assay Details
| Gene Symbol: APOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003369.2 | NP_001003369.2 | 127 | 147 | ||
| GenBank mRNA | DN335595.1 | - | 127 | 147 | ||
| DN757368.1 | - | 91 | 147 | |||
| M17178.1 | - | 73 | 147 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003369.2 transcripts.
Set Membership: |
|

APOC3
Genomic Map