Assay Details
| Gene Symbol: APOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003369.2 | NP_001003369.2 | 254 | 100 | ||
| GenBank mRNA | DN757368.1 | - | 218 | 100 | ||
| M17178.1 | - | 200 | 100 | |||
More Information
Set Membership: |
|

APOC3
Genomic Map