Assay Details
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014768.1 | NP_001014768.1 | 3 - 4 | 694 | 73 | |
GenBank mRNA | LC130496.1 | - | 3 - 4 | 655 | 73 | |
LC130498.1 | - | 3 - 4 | 655 | 73 | ||
LC130499.1 | - | 3 - 4 | 655 | 73 | ||
LC130501.1 | - | 3 - 4 | 655 | 73 | ||
M29611.1 | - | 3 - 4 | 694 | 73 | ||
M57528.1 | - | 3 - 4 | 643 | 73 | ||
M57529.1 | - | 3 - 4 | 643 | 73 | ||
M57531.1 | - | 3 - 4 | 643 | 73 | ||
M57532.1 | - | 3 - 4 | 643 | 73 | ||
M57533.1 | - | 3 - 4 | 643 | 73 | ||
M57534.1 | - | 3 - 4 | 643 | 73 | ||
M57535.1 | - | 3 - 4 | 643 | 73 | ||
M57536.1 | - | 3 - 4 | 643 | 73 | ||
M57537.1 | - | 3 - 4 | 643 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
