Assay Details
| Gene Symbol: RAB5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003317.2 | NP_001003317.1 | 1 - 2 | 30 | 71 | |
| GenBank mRNA | M35520.1 | - | 1 - 2 | 30 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003317.2 transcripts.
Set Membership: |
|

RAB5A
Genomic Map