Assay Details
| Gene Symbol: KCNMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003299.1 | NP_001003299.1 | 1 - 2 | 141 | 61 | |
| XM_005618870.3 | XP_005618927.1 | 2 - 3 | 581 | 61 | ||
| GenBank mRNA | U41002.1 | - | 1 - 2 | 141 | 61 | |
More Information
Set Membership: |
|

KCNMB1
Genomic Map