Assay Details
| Gene Symbol: KLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003262.1 | NP_001003262.1 | 4 - 5 | 634 | 62 | |
| GenBank mRNA | X75479.1 | - | 634 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003262.1 | NP_001003262.1 | 4 - 5 | 634 | 62 | |
| GenBank mRNA | X75479.1 | - | 634 | 62 | ||
Set Membership: |
|