Assay Details
Gene Symbol: ATP6V0C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_537002.6 | XP_537002.1 | 1 - 2 | 235 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
