Assay Details
| Gene Symbol: LOC484343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616238.3 | XP_005616295.1 | 5 - 6 | 1011 | 69 | |
| XM_014114960.2 | XP_013970435.1 | 5 - 6 | 1018 | 69 | ||
| XM_014114961.2 | XP_013970436.1 | 5 - 6 | 1010 | 69 | ||
More Information
Set Membership: |
|

LOC484343
Genomic Map