Assay Details
| Gene Symbol: CDKL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313879.1 | NP_001300808.1 | 8 - 9 | 984 | 117 | |
| XM_022404344.1 | XP_022260052.1 | 9 - 10 | 1114 | 117 | ||
More Information
Set Membership: |
|

CDKL4
Genomic Map