Assay Details
Gene Symbol: FEZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630331.3 | XP_005630388.1 | 8 - 9 | 1148 | 73 | |
XM_005630332.3 | XP_005630389.1 | 7 - 8 | 1067 | 73 | ||
XM_022404627.1 | XP_022260335.1 | 8 - 9 | 1223 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
