Assay Details
| Gene Symbol: BIRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080725.1 | NP_001074194.1 | 7 - 8 | 1615 | 90 | |
| XM_005619513.3 | XP_005619570.1 | 8 - 9 | 3910 | 90 | ||
| XM_005619514.3 | XP_005619571.1 | 8 - 9 | 5009 | 90 | ||
| XM_022418028.1 | XP_022273736.1 | 8 - 9 | 3972 | 90 | ||
| XM_022418029.1 | XP_022273737.1 | 8 - 9 | 3822 | 90 | ||
| GenBank mRNA | EF102104.1 | - | 7 - 8 | 1615 | 90 | |
More Information
Set Membership: |
|

BIRC3
Genomic Map