Assay Details
| Gene Symbol: TUBA4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536077.6 | XP_536077.2 | 1 - 2 | 107 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_536077.6 transcripts.
Set Membership: |
|

TUBA4A
Genomic Map