Assay Details
| Gene Symbol: TUBA4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640689.3 | XP_005640746.1 | 3 - 4 | 600 | 88 | |
| XM_536077.6 | XP_536077.2 | 3 - 4 | 479 | 88 | ||
More Information
Set Membership: |
|

TUBA4A
Genomic Map