Assay Details
| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287127.1 | NP_001274056.1 | 1 - 2 | 92 | 94 | |
| XM_022418020.1 | XP_022273728.1 | 1 - 2 | 167 | 94 | ||
| GenBank mRNA | DN392511.1 | - | 1 - 2 | 92 | 94 | |
More Information
Set Membership: |
|

NIP7
Genomic Map