Assay Details
Gene Symbol: SPAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014106936.2 | XP_013962411.1 | 3 - 4 | 439 | 68 | |
XM_014106939.2 | XP_013962414.1 | 3 - 4 | 375 | 68 | ||
XM_022409107.1 | XP_022264815.1 | 3 - 4 | 697 | 68 | ||
XM_022409108.1 | XP_022264816.1 | 3 - 4 | 715 | 68 | ||
XM_022409109.1 | XP_022264817.1 | 3 - 4 | 750 | 68 | ||
XM_022409110.1 | XP_022264818.1 | 3 - 4 | 561 | 68 | ||
XM_022409111.1 | XP_022264819.1 | 3 - 4 | 705 | 68 | ||
XM_022409112.1 | XP_022264820.1 | 3 - 4 | 717 | 68 | ||
XM_022409113.1 | XP_022264821.1 | 3 - 4 | 753 | 68 | ||
XM_022409114.1 | XP_022264822.1 | 3 - 4 | 751 | 68 | ||
XM_022409115.1 | XP_022264823.1 | 3 - 4 | 480 | 68 | ||
XM_022409116.1 | XP_022264824.1 | 3 - 4 | 754 | 68 | ||
XM_022409117.1 | XP_022264825.1 | 3 - 4 | 732 | 68 | ||
XM_022409118.1 | XP_022264826.1 | 3 - 4 | 725 | 68 | ||
XM_022409119.1 | XP_022264827.1 | 3 - 4 | 735 | 68 | ||
XM_542978.5 | XP_542978.2 | 3 - 4 | 583 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
